siRNA Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001986.2 | NP_001001986.2 | 8 | 2210 | ||
| NM_001134300.2 | NP_001127772.1 | 11 | 2995 | |||
| GenBank mRNA | AK147524.1 | - | 11 | 2997 | ||
| AK164107.1 | - | 1 | 615 | |||
| AK164526.1 | - | 2 | 1167 | |||
| BC029060.1 | - | 1 | 972 | |||
| BC075648.1 | - | 10 | 2405 | 30620653 | ||
| BC079552.1 | - | 8 | 2152 | |||
| BC094234.1 | - | 7 | 2061 | |||

Nol4l
Genomic Map