siRNA Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002004.2 | NP_001002004.2 | 6 | 638 | ||
| GenBank mRNA | AK149719.1 | - | 6 | 609 | ||
| AK150829.1 | - | 6 | 639 | |||
| BC019798.1 | - | 6 | 591 | |||
| BC075633.1 | - | 6 | 598 | 30537398 | ||
| BC080706.1 | - | 6 | 615 | |||

2610507B11Rik
Genomic Map