siRNA Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002004.2 | NP_001002004.2 | 11 | 1308 | ||
| GenBank mRNA | AK149719.1 | - | 11 | 1279 | ||
| BC075633.1 | - | 11 | 1268 | 30537398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610507B11Rik| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002004.2 | NP_001002004.2 | 11 | 1308 | ||
| GenBank mRNA | AK149719.1 | - | 11 | 1279 | ||
| BC075633.1 | - | 11 | 1268 | 30537398 | ||