siRNA Details
| Gene Symbol: Clint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002022.1 | NP_001002022.1 | 8 | 1079 | ||
| XM_006246149.1 | XP_006246211.1 | 8 | 1073 | |||
| XM_008767688.1 | XP_008765910.1 | 7 | 5569 | |||
| XM_008767689.1 | XP_008765911.1 | 8 | 981 | |||
| GenBank mRNA | BC076397.1 | - | 8 | 1079 | 7109965 | |
| Gene Symbol: LOC317516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773226.1 | XP_008771448.1 | 1 | 991 | ||

Clint1
Genomic Map