siRNA Details
| Gene Symbol: Slco4c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002024.1 | NP_001002024.1 | 13 | 2150 | ||
| GenBank mRNA | AF454761.1 | - | 13 | 2150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco4c1| Gene Symbol: Slco4c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002024.1 | NP_001002024.1 | 13 | 2150 | ||
| GenBank mRNA | AF454761.1 | - | 13 | 2150 | ||