siRNA Details
| Gene Symbol: TCAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB018281.1 | - | 9 | 3560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCAF1| Gene Symbol: TCAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB018281.1 | - | 9 | 3560 | ||