siRNA Details
| Gene Symbol: Kprp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002290.1 | NP_001002290.1 | 1 | 2319 | ||
| GenBank mRNA | AF548002.1 | - | 1 | 2319 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kprp| Gene Symbol: Kprp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002290.1 | NP_001002290.1 | 1 | 2319 | ||
| GenBank mRNA | AF548002.1 | - | 1 | 2319 | ||