siRNA Details
| Gene Symbol: KIAA0513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286565.1 | NP_001273494.1 | 13 | 6844 | ||
| NM_001286566.1 | NP_001273495.1 | 12 | 6463 | |||
| NM_014732.3 | NP_055547.1 | 13 | 6486 | |||
| XM_005256265.3 | XP_005256322.1 | 13 | 6535 | |||
| XM_017023912.1 | XP_016879401.1 | 13 | 9061 | |||
| GenBank mRNA | AB011085.1 | - | 13 | 6840 | ||
| AF055005.1 | - | 1 | 930 | |||
| U79288.1 | - | 1 | 677 | |||

KIAA0513
Genomic Map