siRNA Details
Gene Symbol: Cpa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002808.2 | NP_001002808.2 | 9 | 1375 | ||
XM_006236215.2 | XP_006236277.1 | 10 | 1914 | |||
XM_006236216.2 | XP_006236278.1 | 10 | 1638 | |||
XM_006236217.2 | XP_006236279.1 | 10 | 1598 | |||
XM_006236218.2 | XP_006236280.1 | 10 | 1169 | |||
XM_006236219.2 | XP_006236281.1 | 9 | 2063 | |||
XM_008762801.1 | XP_008761023.1 | 10 | 1954 | |||
XM_008762802.1 | XP_008761024.1 | 10 | 1930 | |||
XM_008762803.1 | XP_008761025.1 | 9 | 2063 | |||
GenBank mRNA | BC085840.1 | - | 9 | 1375 | 7135802 |
