siRNA Details
| Gene Symbol: Cpa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002808.2 | NP_001002808.2 | 10 | 1506 | ||
| XM_006236215.2 | XP_006236277.1 | 11 | 2045 | |||
| XM_006236216.2 | XP_006236278.1 | 11 | 1769 | |||
| XM_006236217.2 | XP_006236279.1 | 11 | 1729 | |||
| XM_006236218.2 | XP_006236280.1 | 11 | 1300 | |||
| XM_006236219.2 | XP_006236281.1 | 10 | 2194 | |||
| XM_008762801.1 | XP_008761023.1 | 11 | 2085 | |||
| XM_008762802.1 | XP_008761024.1 | 11 | 2061 | |||
| GenBank mRNA | BC085840.1 | - | 10 | 1506 | 7135802 | |

Cpa5
Genomic Map