siRNA Details
| Gene Symbol: Atf6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002809.3 | NP_001002809.2 | 17 | 2145 | ||
| XM_006255983.1 | XP_006256045.1 | 17 | 2270 | |||
| GenBank mRNA | BC098631.1 | - | 17 | 2136 | 7103893 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf6b| Gene Symbol: Atf6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002809.3 | NP_001002809.2 | 17 | 2145 | ||
| XM_006255983.1 | XP_006256045.1 | 17 | 2270 | |||
| GenBank mRNA | BC098631.1 | - | 17 | 2136 | 7103893 | |