siRNA Details
| Gene Symbol: Ggt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002820.1 | NP_001002820.1 | 4 | 1247 | ||
| XM_008767848.1 | XP_008766070.1 | 3 | 1436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggt6| Gene Symbol: Ggt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002820.1 | NP_001002820.1 | 4 | 1247 | ||
| XM_008767848.1 | XP_008766070.1 | 3 | 1436 | |||