siRNA Details
| Gene Symbol: Notch4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002827.1 | NP_001002827.1 | 4 | 924 | ||
| XM_008772739.1 | XP_008770961.1 | 2 | 1019 | |||
| XM_008772740.1 | XP_008770962.1 | 3 | 927 | |||
| XM_008772741.1 | XP_008770963.1 | 2 | 1019 | |||

Notch4
Genomic Map