siRNA Details
| Gene Symbol: Rasl11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002829.1 | NP_001002829.1 | 2 | 122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasl11a| Gene Symbol: Rasl11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002829.1 | NP_001002829.1 | 2 | 122 | ||