siRNA Details
| Gene Symbol: Smoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002835.1 | NP_001002835.1 | 12, 13 | 1335 | ||
| XM_006240284.2 | XP_006240346.1 | 12, 13 | 1641 | |||
| XM_008764785.1 | XP_008763007.1 | 13, 14 | 1741 | |||
| XM_008764786.1 | XP_008763008.1 | 13, 14 | 1708 | |||
| GenBank mRNA | FQ222878.1 | - | 12, 13 | 1592 | ||

Smoc1
Genomic Map