siRNA Details
Gene Symbol: Prm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002850.1 | NP_001002850.1 | 2 | 142 | ||
GenBank mRNA | BC126070.1 | - | 2 | 226 | 7134211 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002850.1 | NP_001002850.1 | 2 | 142 | ||
GenBank mRNA | BC126070.1 | - | 2 | 226 | 7134211 |