siRNA Details
| Gene Symbol: P2ry13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002853.1 | NP_001002853.1 | 1 | 656 | ||
| XM_008760991.1 | XP_008759213.1 | 2 | 962 | |||
| GenBank mRNA | AY639875.1 | - | 1 | 656 | ||
More Information
Set Membership: |
|

P2ry13
Genomic Map