siRNA Details
| Gene Symbol: Tanc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002854.1 | NP_001002854.1 | 9 | 1195 | ||
| XM_006234204.1 | XP_006234266.1 | 8 | 1114 | |||
| XM_006234205.2 | XP_006234267.1 | 7 | 3350 | |||
| XM_006234207.2 | XP_006234269.1 | 4 | 1312 | |||
| XM_008761838.1 | XP_008760060.1 | 7 | 1528 | |||
| XM_008761839.1 | XP_008760061.1 | 7 | 1814 | |||
| XM_008761840.1 | XP_008760062.1 | 5 | 2446 | |||
| GenBank mRNA | AB098072.1 | - | 9 | 1195 | ||

Tanc1
Genomic Map