siRNA Details
| Gene Symbol: Eml5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003402.2 | NP_001003402.2 | 22 | 3606 | ||
| XM_003750213.3 | XP_003750261.1 | 22 | 3653 | |||
| XM_008764827.1 | XP_008763049.1 | 22 | 3653 | |||
| XM_008764828.1 | XP_008763050.1 | 22 | 3654 | |||
| GenBank mRNA | AY445136.1 | - | 22 | 3614 | ||

Eml5
Genomic Map