siRNA Details
| Gene Symbol: Il22ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003404.1 | NP_001003404.1 | 5 | 1936 | ||
| XM_006227664.2 | XP_006227726.1 | 6 | 1989 | |||
| GenBank mRNA | AJ555485.1 | - | 5 | 1958 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il22ra2| Gene Symbol: Il22ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003404.1 | NP_001003404.1 | 5 | 1936 | ||
| XM_006227664.2 | XP_006227726.1 | 6 | 1989 | |||
| GenBank mRNA | AJ555485.1 | - | 5 | 1958 | ||