siRNA Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014747.2 | NP_055562.2 | 8 | 5650 | ||
| XM_011542479.2 | XP_011540781.1 | 7 | 5288 | |||
| GenBank mRNA | D87074.1 | - | 8 | 5656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIMS3| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014747.2 | NP_055562.2 | 8 | 5650 | ||
| XM_011542479.2 | XP_011540781.1 | 7 | 5288 | |||
| GenBank mRNA | D87074.1 | - | 8 | 5656 | ||