siRNA Details
| Gene Symbol: Pard6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003653.1 | NP_001003653.1 | 3 | 756 | ||
| NM_001003654.1 | NP_001003654.1 | 3 | 788 | |||
| XM_006255490.2 | XP_006255552.1 | 3 | 807 | |||
| GenBank mRNA | AY682586.1 | - | 3 | 788 | ||
| AY682587.1 | - | 3 | 756 | |||
| DQ011518.1 | - | 3 | 724 | |||

Pard6a
Genomic Map