siRNA Details
Gene Symbol: Wfdc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003706.3 | NP_001003706.2 | 2, 3 | 219 | ||
GenBank mRNA | AB101676.1 | - | 2, 3 | 235 | ||
BG661061.1 | - | 2, 3 | 219 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wfdc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003706.3 | NP_001003706.2 | 2, 3 | 219 | ||
GenBank mRNA | AB101676.1 | - | 2, 3 | 235 | ||
BG661061.1 | - | 2, 3 | 219 |