siRNA Details
| Gene Symbol: Clec4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003707.1 | NP_001003707.1 | 4, 5 | 417 | ||
| XM_006237322.2 | XP_006237384.1 | 4, 5 | 599 | |||
| GenBank mRNA | AY339882.1 | - | 4, 5 | 417 | ||
| AY494065.1 | - | 4, 5 | 417 | |||

Clec4d
Genomic Map