siRNA Details
| Gene Symbol: Clec4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003707.1 | NP_001003707.1 | 6 | 553 | ||
| GenBank mRNA | AY339882.1 | - | 6 | 553 | ||
| AY494065.1 | - | 6 | 553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4d| Gene Symbol: Clec4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003707.1 | NP_001003707.1 | 6 | 553 | ||
| GenBank mRNA | AY339882.1 | - | 6 | 553 | ||
| AY494065.1 | - | 6 | 553 | |||