siRNA Details
| Gene Symbol: C5ar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003710.2 | NP_001003710.2 | 1204 | |||
| GenBank mRNA | AY741660.1 | - | 1204 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5ar2| Gene Symbol: C5ar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003710.2 | NP_001003710.2 | 1204 | |||
| GenBank mRNA | AY741660.1 | - | 1204 | |||
Set Membership: |
|