siRNA Details
| Gene Symbol: Jph4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003711.1 | NP_001003711.1 | 3 | 1256 | ||
| XM_008770690.1 | XP_008768912.1 | 4 | 2054 | |||
| GenBank mRNA | AY341260.1 | - | 3 | 1256 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_177049, AK048607, BC052376, BC052722, AY341261, and others.

Jph4
Genomic Map