siRNA Details
| Gene Symbol: Kcnk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003820.1 | NP_001003820.1 | 3 | 1180 | ||
| NM_001134912.1 | NP_001128384.1 | 2 | 1085 | |||
| GenBank mRNA | AY567970.1 | - | 3 | 1180 | ||
| EU152989.1 | - | 2 | 1085 | |||

Kcnk18
Genomic Map