siRNA Details
| Gene Symbol: Slc5a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003915.2 | NP_001003915.1 | 14 | 1914 | ||
| NM_001177624.1 | NP_001171095.1 | 14 | 1902 | |||
| GenBank mRNA | AF529222.1 | - | 14 | 1922 | ||
| AK085356.1 | - | 14 | 1897 | |||
| AK128951.1 | - | 14 | 1894 | |||
| AK144004.1 | - | 14 | 1835 | |||
| AK165419.1 | - | 14 | 1895 | |||
| AY964639.1 | - | 14 | 1605 | |||
| BC121791.1 | - | 14 | 1598 | |||
| BC141255.1 | - | 14 | 1830 | 9055867 | ||

Slc5a12
Genomic Map