siRNA Details
| Gene Symbol: Cyp4x1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003947.1 | NP_001003947.1 | 5, 6 | 631 | ||
| GenBank mRNA | AJ786769.1 | - | 5, 6 | 631 | ||
| AK038526.1 | - | 607 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4x1| Gene Symbol: Cyp4x1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003947.1 | NP_001003947.1 | 5, 6 | 631 | ||
| GenBank mRNA | AJ786769.1 | - | 5, 6 | 631 | ||
| AK038526.1 | - | 607 | ||||