siRNA Details
| Gene Symbol: Dnmt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003959.1 | NP_001003959.1 | 22, 23 | 2730 | ||
| XM_006235340.2 | XP_006235402.1 | 21, 22 | 2603 | |||
| XM_006235342.2 | XP_006235404.1 | 21, 22 | 2726 | |||
| XM_006235347.2 | XP_006235409.1 | 20, 21 | 2541 | |||
| XM_008762364.1 | XP_008760586.1 | 20, 21 | 4005 | |||
| XM_008762365.1 | XP_008760587.1 | 20, 21 | 4005 | |||
| XM_008762366.1 | XP_008760588.1 | 20, 21 | 4002 | |||
| XM_008762367.1 | XP_008760589.1 | 19, 20 | 3945 | |||
| XM_008762368.1 | XP_008760590.1 | 21, 22 | 2383 | |||
| XM_008762369.1 | XP_008760591.1 | 19, 20 | 3942 | |||

Dnmt3b
Genomic Map