siRNA Details
| Gene Symbol: G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003975.3 | NP_001003975.1 | 3 | 1224 | ||
| XM_006256073.2 | XP_006256135.1 | 2 | 1435 | |||
| GenBank mRNA | BC101859.1 | - | 3 | 1204 | 7937508 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

G4| Gene Symbol: G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003975.3 | NP_001003975.1 | 3 | 1224 | ||
| XM_006256073.2 | XP_006256135.1 | 2 | 1435 | |||
| GenBank mRNA | BC101859.1 | - | 3 | 1204 | 7937508 | |