siRNA Details
| Gene Symbol: G6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003976.1 | NP_001003976.1 | 1, 2 | 57 | ||
| XM_006256069.2 | XP_006256131.1 | 1, 2 | 609 | |||
| XM_008772746.1 | XP_008770968.1 | 1, 2 | 609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G6b| Gene Symbol: G6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003976.1 | NP_001003976.1 | 1, 2 | 57 | ||
| XM_006256069.2 | XP_006256131.1 | 1, 2 | 609 | |||
| XM_008772746.1 | XP_008770968.1 | 1, 2 | 609 | |||