siRNA Details
Gene Symbol: Ano7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004071.1 | NP_001004071.1 | 7, 8 | 759 | ||
XM_008767382.1 | XP_008765604.1 | 8, 9 | 2413 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ano7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004071.1 | NP_001004071.1 | 7, 8 | 759 | ||
XM_008767382.1 | XP_008765604.1 | 8, 9 | 2413 |