siRNA Details
| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004072.2 | NP_001004072.2 | 6 | 610 | ||
| GenBank mRNA | BC079369.1 | - | 6 | 602 | ||
| BC098897.1 | - | 6 | 610 | 7103543 | ||
| Z12158.1 | - | 6 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdha1| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004072.2 | NP_001004072.2 | 6 | 610 | ||
| GenBank mRNA | BC079369.1 | - | 6 | 602 | ||
| BC098897.1 | - | 6 | 610 | 7103543 | ||
| Z12158.1 | - | 6 | 629 | |||