siRNA Details
| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004072.2 | NP_001004072.2 | 11 | 1847 | ||
| GenBank mRNA | BC079369.1 | - | 11 | 1839 | ||
| BC098897.1 | - | 11 | 1847 | 7103543 | ||
| FQ214890.1 | - | 1 | 177 | |||
| FQ235262.1 | - | 1 | 104 | |||
| Z12158.1 | - | 11 | 1866 | |||

Pdha1
Genomic Map