siRNA Details
| Gene Symbol: Egln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004083.1 | NP_001004083.1 | 6 | 1508 | ||
| XM_006228574.2 | XP_006228636.1 | 6 | 1518 | |||
| XM_006228575.2 | XP_006228637.1 | 6 | 1557 | |||
| GenBank mRNA | AY952204.1 | - | 5 | 1238 | ||
| BC078907.1 | - | 6 | 1508 | 7113703 | ||
| BC081858.1 | - | 6 | 1531 | 7107517 | ||

Egln2
Genomic Map