siRNA Details
| Gene Symbol: Egln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004083.1 | NP_001004083.1 | 4 | 1365 | ||
| XM_006228574.2 | XP_006228636.1 | 4 | 1375 | |||
| XM_006228575.2 | XP_006228637.1 | 4 | 1414 | |||
| GenBank mRNA | AY952204.1 | - | 3 | 1095 | ||
| BC078907.1 | - | 4 | 1365 | 7113703 | ||
| BC081858.1 | - | 4 | 1388 | 7107517 | ||

Egln2
Genomic Map