siRNA Details
| Gene Symbol: SCRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145513.1 | NP_001138985.1 | 8 | 2372 | ||
| NM_001145514.1 | NP_001138986.1 | 8 | 2304 | |||
| NM_001145515.1 | NP_001138987.1 | 7 | 2204 | |||
| NM_014766.4 | NP_055581.3 | 8 | 2364 | |||
| GenBank mRNA | AB071705.1 | - | 8 | 2254 | ||
| BC040492.1 | - | 8 | 2316 | 5314711 | ||
| D83777.2 | - | 8 | 2307 | |||

SCRN1
Genomic Map