siRNA Details
| Gene Symbol: Txndc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004092.2 | NP_001004092.1 | 4, 5 | 286 | ||
| GenBank mRNA | AY496270.1 | - | 4, 5 | 289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txndc8| Gene Symbol: Txndc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004092.2 | NP_001004092.1 | 4, 5 | 286 | ||
| GenBank mRNA | AY496270.1 | - | 4, 5 | 289 | ||