siRNA Details
| Gene Symbol: Gjb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004099.1 | NP_001004099.1 | 2 | 1652 | ||
| GenBank mRNA | BC078952.1 | - | 2 | 1652 | 7110122 | |
| X51615.1 | - | 2 | 1649 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gjb2| Gene Symbol: Gjb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004099.1 | NP_001004099.1 | 2 | 1652 | ||
| GenBank mRNA | BC078952.1 | - | 2 | 1652 | 7110122 | |
| X51615.1 | - | 2 | 1649 | |||