siRNA Details
| Gene Symbol: KIAA0141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014773.4 | NP_055588.3 | 12 | 1731 | ||
| XM_005268552.2 | XP_005268609.1 | 11 | 1606 | |||
| XM_006714812.3 | XP_006714875.1 | 6 | 1072 | |||
| GenBank mRNA | AK292869.1 | - | 12 | 1680 | ||
| BC007855.2 | - | 12 | 1707 | 4298364 | ||
| BC010452.1 | - | 12 | 1622 | 4155782 | ||
| BC011269.1 | - | 12 | 1622 | 4155755 | ||
| BC011926.1 | - | 12 | 1652 | 4543650 | ||
| BC035486.1 | - | 12 | 1647 | |||
| D50931.1 | - | 12 | 1689 | |||

KIAA0141
Genomic Map