siRNA Details
| Gene Symbol: TUSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004125.2 | NP_001004125.1 | 1 | 1471 | ||
| GenBank mRNA | AY168647.2 | - | 1 | 1053 | ||
| BC017510.1 | - | 1 | 846 | |||
| BC028316.1 | - | 1 | 570 | |||
| BC036931.1 | - | 1 | 550 | |||
| BC046919.1 | - | 1 | 912 | |||
| BC110824.1 | - | 1 | 1053 | 5169320 | ||

TUSC1
Genomic Map