siRNA Details
| Gene Symbol: TUSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004125.2 | NP_001004125.1 | 1 | 1463 | ||
| GenBank mRNA | AY168647.2 | - | 1 | 1045 | ||
| BC017510.1 | - | 1 | 838 | |||
| BC028316.1 | - | 1 | 562 | |||
| BC036931.1 | - | 1 | 542 | |||
| BC046919.1 | - | 1 | 904 | |||
| BC110824.1 | - | 1 | 1045 | 5169320 | ||

TUSC1
Genomic Map