siRNA Details
| Gene Symbol: Stfa2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004129.1 | NP_001004129.1 | 2 | 144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stfa2l1| Gene Symbol: Stfa2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004129.1 | NP_001004129.1 | 2 | 144 | ||