siRNA Details
| Gene Symbol: Nlrp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004142.2 | NP_001004142.2 | 3 | 392 | ||
| GenBank mRNA | AK157034.1 | - | 3 | 393 | ||
| AY355339.1 | - | 2 | 174 | |||
| DQ117601.1 | - | 2 | 333 | |||
| KC539856.1 | - | 2 | 174 | |||
| KC539857.1 | - | 2 | 174 | |||
| KC539858.1 | - | 2 | 174 | |||
| KC539859.1 | - | 2 | 174 | |||
| KC539860.1 | - | 2 | 174 | |||
| KC539861.1 | - | 2 | 174 | |||

Nlrp1a
Genomic Map