siRNA Details
| Gene Symbol: AU018091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004153.2 | NP_001004153.2 | 12 | 3270 | ||
| GenBank mRNA | AB201954.1 | - | 12 | 3221 | ||
| BC050126.1 | - | 12 | 3148 | 30024412 | ||
| BC080735.1 | - | 12 | 3270 | 30112559 | ||
| BC086454.1 | - | 10 | 3190 | 30460703 | ||

AU018091
Genomic Map