siRNA Details
| Gene Symbol: 9930012K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004155.2 | NP_001004155.2 | 7 | 1837 | ||
| NM_001112735.1 | NP_001106206.1 | 7 | 1834 | |||
| GenBank mRNA | AK036801.1 | - | 2 | 1793 | ||
| AK132259.1 | - | 7 | 1811 | |||
| BC032212.1 | - | 7 | 1777 | |||
| BC080738.1 | - | 7 | 1817 | 30433488 | ||

9930012K11Rik
Genomic Map