siRNA Details
Gene Symbol: Gm5142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004158.2 | NP_001004158.1 | 2 | 482 | ||
GenBank mRNA | AK076969.1 | - | 2 | 483 | ||
BC099536.1 | - | 2 | 454 | 30918677 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004158.2 | NP_001004158.1 | 2 | 482 | ||
GenBank mRNA | AK076969.1 | - | 2 | 483 | ||
BC099536.1 | - | 2 | 454 | 30918677 |