siRNA Details
Gene Symbol: Gm1553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255990.1 | NP_001242919.1 | 3 | 311 | ||
GenBank mRNA | AY702028.1 | - | 3 | 311 | ||
BC141250.1 | - | 3 | 301 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm1553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255990.1 | NP_001242919.1 | 3 | 311 | ||
GenBank mRNA | AY702028.1 | - | 3 | 311 | ||
BC141250.1 | - | 3 | 301 |